Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133766
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
13
2362
NM_001162952.2
NP_001156424.1
5
892
NM_001162953.2
NP_001156425.1
13
2365
NM_001289608.1
NP_001276537.1
14
2171
NM_001289609.1
NP_001276538.1
6
1005
NM_001289610.1
NP_001276539.1
6
576
NM_032943.4
NP_116561.1
14
2541
NM_206929.3
NP_996812.1
7
696
NM_206930.3
NP_996813.1
4
869
XM_005274057.1
XP_005274114.1
13
6001
XM_005274060.1
XP_005274117.1
14
2185
XM_005274061.1
XP_005274118.1
14
2182
XM_005274062.2
XP_005274119.1
15
2606
XM_005274063.1
XP_005274120.1
13
2065
XM_005274066.1
XP_005274123.1
12
2014
XM_005274067.4
XP_005274124.1
6
2308
XM_011545105.1
XP_011543407.1
14
6049
XM_011545106.1
XP_011543408.1
14
6046
XM_011545107.2
XP_011543409.1
15
6470
XM_011545108.1
XP_011543410.1
13
5929
XM_011545109.2
XP_011543411.1
14
6031
XM_011545110.1
XP_011543412.1
14
6049
XM_011545114.2
XP_011543416.1
5
2214
XM_011545115.1
XP_011543417.1
3
308
XM_011545116.2
XP_011543418.1
4
2093
XM_017017933.1
XP_016873422.1
13
5998
XM_017017934.1
XP_016873423.1
15
6467
XM_017017935.1
XP_016873424.1
11
5845
XM_017017936.1
XP_016873425.1
11
5543
XM_017017937.1
XP_016873426.1
14
2086
XM_017017938.1
XP_016873427.1
13
2044
XM_017017939.1
XP_016873428.1
7
2352
XM_017017940.1
XP_016873429.1
6
2304
GenBank mRNA
AB046817.1
-
14
2552
AJ303364.1
-
2
188
AK024872.1
-
4
1033
AK074737.1
-
7
653
AK092121.1
-
4
687
AK124754.1
-
7
2277
AK127504.1
-
8
3240
AK131365.1
-
7
1169
AK222707.1
-
4
1033
AK222710.1
-
4
1033
AK296224.1
-
14
2153
AK296740.1
-
11
2101
AK298604.1
-
6
494
AL601023.1
-
6
549
AY386362.1
-
7
4556
BC015540.2
-
4
363
BC068495.1
-
6
730
BC110315.1
-
5
421
BC136450.1
-
13
2297
9020439
BC144114.1
-
13
2297
9052634
BC144115.1
-
13
2294
9052635
BX647671.1
-
8
2992
CB046277.1
-
7
653
EU035829.1
-
13
2018
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0