Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037174.1 | NP_001032251.1 | 6 | 594 | ||
NM_012097.3 | NP_036229.1 | 6 | 850 | |||
NM_177985.2 | NP_817114.2 | 6 | 592 | |||
GenBank mRNA | AF087889.1 | - | 780 | |||
AF100740.1 | - | 6 | 597 | |||
AK292714.1 | - | 6 | 760 | |||
AK298937.1 | - | 6 | 538 | |||
BC001254.1 | - | 6 | 768 | 3453177 | ||
BX461356.2 | - | 6 | 565 |