Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282062.1 | NP_001268991.1 | 6 | 684 | ||
NM_001282063.1 | NP_001268992.1 | 6 | 646 | |||
NM_001282068.1 | NP_001268997.1 | 5 | 611 | |||
NM_003586.2 | NP_003577.2 | 5 | 645 | |||
NR_104089.1 | 5 | 436 | ||||
NR_104090.1 | 5 | 436 | ||||
XM_011545975.1 | XP_011544277.1 | 7 | 1070 | |||
XM_017023776.1 | XP_016879265.1 | 9 | 1288 | |||
XM_017023777.1 | XP_016879266.1 | 6 | 1225 | |||
XM_017023778.1 | XP_016879267.1 | 5 | 699 | |||
GenBank mRNA | AK293651.1 | - | 5 | 385 | ||
BC041769.1 | - | 6 | 647 | 4819032 | ||
BC055284.1 | - | 5 | 645 | 5767656 | ||
BC063436.1 | - | 6 | 610 | 5394616 | ||
BI757951.1 | - | 7 | 556 | |||
BX460259.2 | - | 5 | 365 | |||
D31897.1 | - | 5 | 566 |