Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012138.3 | NP_036270.1 | 11 | 1825 | ||
GenBank mRNA | AF083208.1 | - | 11 | 1742 | ||
AF161395.1 | - | 6 | 495 | |||
AJ249940.2 | - | 11 | 1680 | |||
AK026173.1 | - | 9 | 1251 | |||
AK057229.1 | - | 11 | 1825 | |||
BC000591.2 | - | 11 | 1705 | 3346769 |