Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193534.1 | NP_001180463.1 | 2 | 247 | ||
NM_012161.3 | NP_036293.1 | 2 | 247 | |||
NR_036464.1 | 2 | 247 | ||||
XM_006713959.3 | XP_006714022.1 | 3 | 343 | |||
XM_011513832.2 | XP_011512134.1 | 2 | 176 | |||
GenBank mRNA | AF142481.1 | - | 2 | 169 | ||
AF157323.1 | - | 2 | 108 | |||
AF176700.1 | - | 2 | 131 | |||
AF199420.1 | - | 2 | 136 | |||
AK310860.1 | - | 2 | 176 | |||
BC012320.1 | - | 2 | 176 | |||
BC030656.1 | - | 2 | 218 | 4837080 | ||
BI464829.1 | - | 2 | 226 | |||
DB080171.1 | - | 2 | 248 |