Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 3, 4 | 679 | ||
XM_005274604.2 | XP_005274661.1 | 3, 4 | 679 | |||
XM_005274606.3 | XP_005274663.1 | 4, 5 | 821 | |||
XM_011545592.2 | XP_011543894.1 | 4, 5 | 650 | |||
XM_011545594.2 | XP_011543896.1 | 2, 3 | 100 | |||
XM_011545595.1 | XP_011543897.1 | 2, 3 | 110 | |||
XM_011545597.1 | XP_011543899.1 | 3, 4 | 610 | |||
XM_017029909.1 | XP_016885398.1 | 2, 3 | 130 | |||
XM_017029910.1 | XP_016885399.1 | 3, 4 | 338 | |||
GenBank mRNA | AK225847.1 | - | 3, 4 | 610 | ||
AK289677.1 | - | 3, 4 | 660 | |||
AK290354.1 | - | 3, 4 | 664 | |||
BC042830.1 | - | 3, 4 | 610 | |||
BC052809.1 | - | 3, 4 | 536 | |||
BC062432.1 | - | 3, 4 | 649 | |||
BC096344.3 | - | 3, 4 | 494 | 40008704 | ||
BC096345.1 | - | 3, 4 | 494 | |||
GU727634.1 | - | 2, 3 | 320 | |||
Y15164.1 | - | 3, 4 | 631 | |||
Y16355.1 | - | 3, 4 | 631 |