Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135004.2 | NP_001128476.2 | 5 | 1973 | ||
NM_001135005.2 | NP_001128477.1 | 4 | 2180 | |||
NM_012266.5 | NP_036398.3 | 4 | 1828 | |||
XM_005251428.3 | XP_005251485.1 | 4 | 1747 | |||
XM_006716751.2 | XP_006716814.1 | 5 | 1837 | |||
XM_011517842.2 | XP_011516144.1 | 5 | 2414 | |||
GenBank mRNA | AF087870.1 | - | 5 | 1830 | ||
AK023253.1 | - | 4 | 1724 |