Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 5 | 649 | ||
NM_001311256.1 | NP_001298185.1 | 4 | 561 | |||
NM_012388.3 | NP_036520.1 | 5 | 815 | |||
NR_132350.1 | 6 | 823 | ||||
NR_132351.1 | 5 | 811 | ||||
NR_132352.1 | 4 | 727 | ||||
NR_132353.1 | 4 | 673 | ||||
NR_132354.1 | 4 | 669 | ||||
NR_132355.1 | 3 | 585 | ||||
NR_132356.1 | 5 | 517 | ||||
NR_132357.1 | 4 | 430 | ||||
NR_132358.1 | 3 | 419 | ||||
NR_132359.1 | 3 | 287 | ||||
GenBank mRNA | AF080470.1 | - | 5 | 719 | ||
AK057545.1 | - | 5 | 555 | |||
AK128626.1 | - | 6 | 792 | |||
AK315581.1 | - | 5 | 533 | |||
AL596952.1 | - | 5 | 588 | |||
BC004819.1 | - | 5 | 536 | 3450397 | ||
DA226593.1 | - | 3 | 357 | |||
DA230439.1 | - | 3 | 420 | |||
DA233328.1 | - | 3 | 276 | |||
DA237963.1 | - | 3 | 288 | |||
DA247079.1 | - | 4 | 447 | |||
DA523523.1 | - | 4 | 375 | |||
DA798595.1 | - | 4 | 441 | |||
DA859617.1 | - | 4 | 378 | |||
DB287075.1 | - | 3 | 358 | |||
DB289156.1 | - | 5 | 460 |