Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 5 | 3520 | ||
NM_001311256.1 | NP_001298185.1 | 4 | 3432 | |||
NM_012388.3 | NP_036520.1 | 5 | 3686 | |||
NR_132350.1 | 6 | 3694 | ||||
NR_132351.1 | 5 | 3682 | ||||
NR_132352.1 | 4 | 3598 | ||||
NR_132353.1 | 4 | 3544 | ||||
NR_132354.1 | 4 | 3540 | ||||
NR_132355.1 | 3 | 3456 | ||||
NR_132356.1 | 5 | 3388 | ||||
NR_132357.1 | 4 | 3301 | ||||
NR_132358.1 | 3 | 3290 | ||||
NR_132359.1 | 3 | 3158 | ||||
GenBank mRNA | AF131859.1 | - | 1 | 1436 | ||
AK128626.1 | - | 6 | 3663 | |||
BP423071.1 | - | 19 |