Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 17 | 1890 | ||
NM_001256733.1 | NP_001243662.1 | 16 | 1806 | |||
NM_001256734.1 | NP_001243663.1 | 16 | 1800 | |||
NM_001256735.1 | NP_001243664.1 | 16 | 1776 | |||
NM_001256736.1 | NP_001243665.1 | 15 | 1677 | |||
NM_012446.3 | NP_036578.2 | 17 | 1866 | |||
XM_011543297.2 | XP_011541599.1 | 16 | 1706 | |||
XM_017009304.1 | XP_016864793.1 | 17 | 1708 | |||
XM_017009305.1 | XP_016864794.1 | 18 | 1797 | |||
XM_017009306.1 | XP_016864795.1 | 16 | 1682 | |||
XM_017009307.1 | XP_016864796.1 | 16 | 1594 | |||
XM_017009308.1 | XP_016864797.1 | 18 | 1969 | |||
XM_017009309.1 | XP_016864798.1 | 17 | 2033 | |||
XM_017009310.1 | XP_016864799.1 | 17 | 2181 | |||
GenBank mRNA | AK294864.1 | - | 15 | 1472 |