Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 3, 4 | 475 | ||
NM_001303041.1 | NP_001289970.1 | 2, 3 | 363 | |||
NM_003637.4 | NP_003628.2 | 6, 7 | 792 | |||
XM_005277436.3 | XP_005277493.1 | 5, 6 | 543 | |||
XM_011510083.2 | XP_011508385.1 | 2, 3 | 223 | |||
XM_017002622.1 | XP_016858111.1 | 4, 5 | 749 | |||
XM_017002623.1 | XP_016858112.1 | 4, 5 | 749 | |||
XM_017002624.1 | XP_016858113.1 | 4, 5 | 749 | |||
XM_017002625.1 | XP_016858114.1 | 4, 5 | 749 | |||
XM_017002627.1 | XP_016858116.1 | 4, 5 | 749 | |||
XM_017002628.1 | XP_016858117.1 | 1 | 204 | |||
GenBank mRNA | AF074015.1 | - | 6, 7 | 638 | ||
AF112345.1 | - | 6, 7 | 693 | |||
AK304155.1 | - | 3, 4 | 331 | |||
AK304183.1 | - | 5, 6 | 622 | |||
AK314255.1 | - | 6, 7 | 641 | |||
BC140831.1 | - | 2, 3 | 238 | 9021699 | ||
BC144637.1 | - | 2, 3 | 238 | 9053168 |