Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013937.3 | NP_039225.1 | 1 | 834 | ||
GenBank mRNA | AJ459857.1 | - | 1 | 834 | ||
BC131608.1 | - | 1 | 815 | 40114992 |
Set Membership: |
G-protein coupled receptor |