Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014035.2 | NP_054754.1 | 7 | 786 | ||
XM_017009394.1 | XP_016864883.1 | 7 | 611 | |||
GenBank mRNA | AF139461.1 | - | 7 | 675 | ||
AK021473.1 | - | 1 | 1252 | |||
BC010886.1 | - | 7 | 746 | 3538426 | ||
BC069012.1 | - | 7 | 809 | 4585781 | ||
BQ278046.1 | - | 7 | 758 |