Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291030.1 | - | 4, 5 | 554 | ||
AL834518.1 | - | 4, 5 | 559 | |||
AY358399.1 | - | 4, 5 | 954 | |||
BC064901.1 | - | 1, 2 | 113 | |||
BC113714.1 | - | 4, 5 | 436 | 8322766 | ||
BC113716.1 | - | 4, 5 | 436 | 8322768 | ||
BX161500.1 | - | 4, 5 | 882 |