Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014050.3 | NP_054769.1 | 4 | 334 | ||
NM_172177.3 | NP_751917.1 | 4 | 337 | |||
NR_038159.1 | 5 | 357 | ||||
NR_038160.1 | 5 | 354 | ||||
NR_038161.1 | 5 | 439 | ||||
GenBank mRNA | AF078860.1 | - | 4 | 316 | ||
AF135160.1 | - | 4 | 320 | |||
AF151038.1 | - | 4 | 308 | |||
AK000285.1 | - | 4 | 310 | |||
AL136659.1 | - | 4 | 311 | |||
BC040240.2 | - | 3 | 273 | 3445213 | ||
BM147739.1 | - | 4 | 335 | |||
BM726733.1 | - | 5 | 312 | |||
BU561400.1 | - | 333 | ||||
CD385500.1 | - | 4 | 330 | |||
CR533495.1 | - | 3 | 175 | |||
DB195199.1 | - | 5 | 415 |