Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014169.3 | NP_054888.2 | 6 | 1136 | ||
GenBank mRNA | AF161483.1 | - | 6 | 793 | ||
AF212243.1 | - | 6 | 766 | |||
BC010893.1 | - | 6 | 773 | |||
BC113533.1 | - | 6 | 1136 | 8322585 | ||
BC113535.1 | - | 6 | 1136 | 8322587 |