Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326380.1 | NP_001313309.1 | 3 | 436 | ||
NM_014320.2 | NP_055135.1 | 3 | 667 | |||
GenBank mRNA | AF117616.1 | - | 3 | 645 | ||
AF411610.1 | - | 3 | 307 | |||
AK057928.1 | - | 3 | 582 | |||
AY427823.1 | - | 3 | 549 | |||
BC008205.1 | - | 3 | 531 | 3869026 | ||
BC010290.1 | - | 3 | 544 | 3445763 | ||
BC016806.1 | - | 3 | 647 | |||
BC016807.1 | - | 3 | 647 | |||
BC093037.1 | - | 3 | 530 | 30333215 |