Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306151.1 | NP_001293080.1 | 10 | 1211 | ||
NM_014395.2 | NP_055210.2 | 9 | 1158 | |||
XM_011531842.1 | XP_011530144.1 | 10 | 1193 | |||
XM_011531843.2 | XP_011530145.1 | 7 | 947 | |||
GenBank mRNA | AF161551.1 | - | 9 | 1134 | ||
AF163254.1 | - | 9 | 1139 | |||
AF178987.1 | - | 9 | 1113 | |||
AF186022.1 | - | 10 | 1208 | |||
AL713793.1 | - | 2 | 1568 | |||
BC012924.1 | - | 9 | 1083 | 4517170 |