Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014399.3 | NP_055214.1 | 6 | 1541 | ||
GenBank mRNA | AF100759.1 | - | 6 | 1462 | ||
AF120265.1 | - | 6 | 1368 | |||
AK026587.1 | - | 6 | 1505 | |||
AK075475.1 | - | 6 | 1486 | |||
AK093487.1 | - | 6 | 1503 | |||
AY358995.1 | - | 6 | 1472 | |||
BC033863.1 | - | 6 | 1391 | 5187772 | ||
BX647338.1 | - | 1 | 1665 | |||
CB852023.1 | - | 1 | 163 |