Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142885.1 | NP_001136357.1 | 9 | 1555 | ||
NM_014548.3 | NP_055363.1 | 10 | 1663 | |||
XM_017022089.1 | XP_016877578.1 | 9 | 1482 | |||
GenBank mRNA | AF177169.1 | - | 10 | 1598 | ||
AK293823.1 | - | 9 | 1443 | |||
BC036184.1 | - | 11 | 1709 | 4815047 | ||
BC064961.1 | - | 10 | 1611 | 6141541 |