Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 9 | 1744 | ||
NM_001034838.2 | NP_001030010.1 | 8 | 1591 | |||
NM_001278339.1 | NP_001265268.1 | 8 | 1786 | |||
NM_001278340.1 | NP_001265269.1 | 9 | 1873 | |||
NM_014592.3 | NP_055407.1 | 8 | 1711 | |||
XM_017009407.1 | XP_016864896.1 | 9 | 1715 | |||
GenBank mRNA | AK074390.1 | - | 8 | 1307 | ||
AY170821.1 | - | 8 | 1387 | |||
AY780424.1 | - | 9 | 1549 | |||
BC035032.1 | - | 8 | 1716 | |||
BC050375.1 | - | 8 | 1478 | 5752207 |