Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146326.1 | NP_001139798.1 | 12 | 1554 | ||
NM_001146327.1 | NP_001139799.1 | 12 | 1641 | |||
NM_015363.4 | NP_056178.3 | 11 | 1478 | |||
GenBank mRNA | AF166122.1 | - | 11 | 1461 | ||
BC104876.1 | - | 11 | 1385 | 8143879 | ||
BC104878.1 | - | 11 | 1469 | 8143881 |