Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134469
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
6
2689
NM_001270727.1
NP_001257656.1
5
2545
NM_001270728.1
NP_001257657.1
4
2525
NM_001270729.1
NP_001257658.1
6
2703
NM_001270730.1
NP_001257659.1
5
2532
NM_001270731.1
NP_001257660.1
6
2790
NM_001270732.1
NP_001257661.1
5
2633
NM_001270733.1
NP_001257662.1
2
2075
NM_001270734.1
NP_001257663.1
5
2646
NM_001270735.1
NP_001257664.1
4
2489
NM_015367.3
NP_056182.2
7
2860
NR_073068.1
5
2458
NR_073069.1
5
2441
XM_011546119.1
XP_011544421.1
7
3223
XM_011546120.1
XP_011544422.1
7
3225
XM_011546121.2
XP_011544423.1
8
3208
XM_011546123.1
XP_011544425.1
6
2588
XM_017028725.1
XP_016884214.1
6
3079
XM_017028726.1
XP_016884215.1
9
3509
XM_017028727.1
XP_016884216.1
7
2658
XM_017028728.1
XP_016884217.1
7
3237
XM_017028729.1
XP_016884218.1
7
2732
XM_017028730.1
XP_016884219.1
5
2446
XM_017028731.1
XP_016884220.1
6
3066
XM_017028732.1
XP_016884221.1
4
2309
XM_017028734.1
XP_016884223.1
6
3167
GenBank mRNA
AF146568.1
-
4
2280
AK027443.1
-
1
1336
AK056248.1
-
6
2547
AK075044.1
-
1
1185
AL133029.1
-
2
1962
AL831982.2
-
5
2420
BC003032.1
-
3
2120
BC007658.2
-
7
2619
3343994
BC016037.1
-
1
1406
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0