Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321389.1 | NP_001308318.1 | 5 | 760 | ||
NM_001321390.1 | NP_001308319.1 | 5 | 405 | |||
NM_001321391.1 | NP_001308320.1 | 5 | 439 | |||
NM_015680.5 | NP_056495.3 | 4 | 497 | |||
NR_135628.1 | 5 | 439 | ||||
NR_135629.1 | 4 | 497 | ||||
GenBank mRNA | AF151815.1 | - | 5 | 664 | ||
AK075274.1 | - | 4 | 453 | |||
AK314975.1 | - | 4 | 440 | |||
AY007160.1 | - | 4 | 470 | |||
BC001393.2 | - | 4 | 420 | 3051397 | ||
BC012821.2 | - | 4 | 469 | 2958621 | ||
BI912613.1 | - | 5 | 406 | |||
CR457401.1 | - | 4 | 332 | |||
CX166359.1 | - | 5 | 756 | |||
DB477110.1 | - | 4 | 484 | |||
HY106884.1 | - | 4 | 497 |