Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289987.1 | NP_001276916.1 | 3 | 753 | ||
NM_001300866.1 | NP_001287795.1 | 2 | 587 | |||
NM_015687.3 | NP_056502.1 | 2 | 587 | |||
XM_005248713.3 | XP_005248770.1 | 2 | 558 | |||
XM_005248715.4 | XP_005248772.1 | 2 | 558 | |||
GenBank mRNA | AB086011.1 | - | 1 | 171 | ||
AK097021.1 | - | 2 | 427 | |||
AK290232.1 | - | 2 | 299 | |||
AL832009.2 | - | 2 | 387 | |||
BC136443.1 | - | 2 | 342 | 9020432 | ||
BC136444.1 | - | 3 | 508 | 9020433 | ||
BX647178.1 | - | 2 | 538 |