Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195303.2 | NP_001182232.1 | 3 | 704 | ||
NM_001254748.1 | NP_001241677.1 | 6 | 762 | |||
NM_001254749.1 | NP_001241678.1 | 5 | 889 | |||
NM_003617.3 | NP_003608.1 | 5 | 877 | |||
GenBank mRNA | AB008109.1 | - | 5 | 620 | ||
AF030108.1 | - | 5 | 615 | |||
AF159570.1 | - | 5 | 583 | |||
BC030059.1 | - | 5 | 602 | 4098248 | ||
BU172751.1 | - | 5 | 619 | |||
BX537427.1 | - | 5 | 621 |