Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015918.3 | NP_057002.2 | 2 | 108 | ||
NM_198202.1 | NP_937845.1 | 2 | 112 | |||
XM_011538441.1 | XP_011536743.1 | 2 | 166 | |||
GenBank mRNA | AF070660.1 | - | 2 | 95 | ||
AF117232.1 | - | 2 | 95 | |||
AJ306296.1 | - | 2 | 95 | |||
AK223206.1 | - | 2 | 80 | |||
AK303144.1 | - | 2 | 93 | |||
BC012505.1 | - | 2 | 95 | 4708484 | ||
BI549018.1 | - | 2 | 117 | |||
BI668578.1 | - | 2 | 116 | |||
BU155588.1 | - | 101 | ||||
CB995722.1 | - | 2 | 111 |