Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015995.3 | NP_057079.2 | 2 | 1494 | ||
GenBank mRNA | AL390127.1 | - | 1 | 219 |