Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282168.1 | NP_001269097.1 | 2 | 200 | ||
NM_001282169.1 | NP_001269098.1 | 2 | 361 | |||
NM_016075.3 | NP_057159.2 | 2 | 227 | |||
GenBank mRNA | AF151903.1 | - | 2 | 148 | ||
AI221133.1 | - | 2 | 361 | |||
AK289740.1 | - | 2 | 170 | |||
BC037279.1 | - | 2 | 176 | 4814999 | ||
BC050439.1 | - | 2 | 150 | 5764280 | ||
BG400860.1 | - | 2 | 178 | |||
BX647847.1 | - | 2 | 168 | |||
CB959894.1 | - | 2 | 184 | |||
CR933653.1 | - | 2 | 315 | |||
DA461148.1 | - | 2 | 228 |