Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282168.1 | NP_001269097.1 | 13 | 1067 | ||
NM_001282169.1 | NP_001269098.1 | 13 | 1228 | |||
NM_016075.3 | NP_057159.2 | 13 | 1094 | |||
GenBank mRNA | AB208956.1 | - | 7 | 3768 | ||
AF151903.1 | - | 13 | 1015 | |||
AK023182.1 | - | 11 | 811 | |||
AK289740.1 | - | 13 | 1037 | |||
BC037279.1 | - | 13 | 1043 | 4814999 | ||
BC050439.1 | - | 13 | 1017 | 5764280 | ||
BX647847.1 | - | 13 | 1035 | |||
CR933653.1 | - | 13 | 1182 |