Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305134.1 | NP_001292063.1 | 10 | 1635 | ||
NM_016192.3 | NP_057276.2 | 10 | 1578 | |||
XM_011510890.2 | XP_011509192.1 | 9 | 1546 | |||
XM_017003739.1 | XP_016859228.1 | 9 | 1615 | |||
GenBank mRNA | AB017269.1 | - | 10 | 1536 | ||
AF179274.2 | - | 10 | 1563 | |||
AF242222.1 | - | 10 | 1578 | |||
AK074507.1 | - | 10 | 1192 | |||
AK074632.1 | - | 10 | 1561 | |||
AK222691.1 | - | 10 | 1583 | |||
AL157430.1 | - | 10 | 1618 | |||
AY358907.1 | - | 10 | 1544 | |||
BC008973.1 | - | 10 | 1519 | 4180709 |