Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282396.1 | NP_001269325.1 | 5 | 1285 | ||
NM_001282397.1 | NP_001269326.1 | 3 | 963 | |||
NM_016257.3 | NP_057341.1 | 4 | 1404 | |||
XM_017001447.1 | XP_016856936.1 | 4 | 1017 | |||
GenBank mRNA | AB001105.1 | - | 4 | 1386 | ||
AK294412.1 | - | 5 | 1100 | |||
AK294813.1 | - | 3 | 945 | |||
AL136591.1 | - | 4 | 1111 | |||
BC030827.1 | - | 4 | 1109 | 4811834 |