Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016306.5 | NP_057390.1 | 8 | 974 | ||
GenBank mRNA | AB028859.2 | - | 8 | 917 | ||
AF228505.1 | - | 8 | 843 | |||
AF277317.1 | - | 8 | 934 | |||
AJ250137.1 | - | 8 | 772 | |||
AK075300.1 | - | 8 | 931 | |||
AK075430.1 | - | 8 | 950 | |||
AK124289.1 | - | 4 | 3370 | |||
AK301292.1 | - | 6 | 633 | |||
AY359043.1 | - | 8 | 910 | |||
BC001144.1 | - | 8 | 912 | 3534756 | ||
BC046500.1 | - | 9 | 1681 | |||
BT007063.1 | - | 8 | 752 | |||
CR457096.1 | - | 8 | 752 |