Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: IPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134779.1 | NP_001128251.1 | 30 | 3303 | ||
NM_016338.4 | NP_057422.3 | 30 | 3243 | |||
GenBank mRNA | AK001696.1 | - | 30 | 3225 | ||
AK023177.1 | - | 18 | 1815 | |||
AK124969.1 | - | 1 | 1050 | |||
AK302781.1 | - | 30 | 3303 | |||
AL162083.1 | - | 4 | 608 | |||
BC031694.1 | - | 22 | 2304 | |||
BC033776.1 | - | 30 | 3205 | 4525238 |
Gene Symbol: IPO11-LRRC70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073584.1 | 4 | 672 |