Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198976.2 | NP_945327.2 | 3 | 326 | ||
GenBank mRNA | AF161479.1 | - | 3 | 258 | ||
AJ238379.1 | - | 3 | 260 | |||
AK001316.1 | - | 3 | 234 | |||
AK023310.1 | - | 3 | 251 | |||
AK023927.1 | - | 3 | 251 | |||
AK293410.1 | - | 3 | 278 | |||
AK304310.1 | - | 3 | 278 | |||
BC014952.1 | - | 3 | 262 | 4860894 | ||
BI459250.1 | - | 3 | 287 | |||
BX647417.1 | - | 3 | 261 | |||
HY015074.1 | - | 3 | 327 |