Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 10 | 1258 | ||
XM_006722284.2 | XP_006722347.1 | 8 | 992 | |||
XM_011523580.1 | XP_011521882.1 | 9 | 1124 | |||
XM_011523581.2 | XP_011521883.1 | 9 | 2861 | |||
GenBank mRNA | AF233395.1 | - | 10 | 1240 | ||
AK002027.1 | - | 8 | 2038 | |||
AK094326.1 | - | 10 | 1410 | |||
AK131437.1 | - | 6 | 2441 | |||
AK131484.1 | - | 4 | 2972 | |||
AK290265.1 | - | 10 | 1226 | |||
AL137626.1 | - | 7 | 826 | |||
BC017305.2 | - | 10 | 1222 | 5087554 | ||
BC101791.1 | - | 10 | 1239 | 8069297 | ||
BC101793.1 | - | 10 | 1238 | 8069299 |