Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135087
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
7
1465
NM_001282780.1
NP_001269709.1
5
775
NM_001282781.1
NP_001269710.1
5
775
NM_001308375.1
NP_001295304.1
5
1014
NM_016552.3
NP_057636.2
6
1302
NM_017844.3
NP_060314.2
5
775
XM_011511300.2
XP_011509602.1
7
1649
XM_011511301.1
XP_011509603.1
7
1649
XM_011511304.2
XP_011509606.1
7
1649
XM_011511305.2
XP_011509607.1
7
1649
XM_011511309.2
XP_011509611.1
7
1649
XM_011511313.2
XP_011509615.1
7
1649
XM_011511315.2
XP_011509617.1
7
1649
XM_011511317.1
XP_011509619.1
5
959
XM_011511318.1
XP_011509620.1
5
959
XM_011511319.2
XP_011509621.1
7
1649
XM_011511322.1
XP_011509624.1
5
959
XM_017004271.1
XP_016859760.1
7
1649
XM_017004272.1
XP_016859761.1
7
1649
XM_017004273.1
XP_016859762.1
7
1649
XM_017004274.1
XP_016859763.1
7
1649
XM_017004275.1
XP_016859764.1
7
1415
XM_017004276.1
XP_016859765.1
8
1736
XM_017004277.1
XP_016859766.1
7
1649
XM_017004278.1
XP_016859767.1
7
1649
XM_017004279.1
XP_016859768.1
7
1649
XM_017004280.1
XP_016859769.1
7
1649
XM_017004281.1
XP_016859770.1
6
1255
XM_017004282.1
XP_016859771.1
6
1499
XM_017004283.1
XP_016859772.1
8
1736
XM_017004284.1
XP_016859773.1
5
959
XM_017004285.1
XP_016859774.1
5
725
XM_017004286.1
XP_016859775.1
6
1499
XM_017004287.1
XP_016859776.1
6
1499
XM_017004288.1
XP_016859777.1
4
809
XM_017004289.1
XP_016859778.1
6
1499
XM_017004290.1
XP_016859779.1
5
1026
XM_017004291.1
XP_016859780.1
7
1469
GenBank mRNA
AB034636.1
-
6
1232
AK000506.1
-
5
742
AK056511.2
-
2
499
AK124879.1
-
1
1824
AK125881.1
-
2
2784
AK126637.1
-
1
2276
AK225534.1
-
5
742
AK296714.1
-
4
528
AK314534.1
-
7
1449
AL136618.1
-
4
521
AL833866.1
-
3
382
BC033495.1
-
5
775
BC073146.1
-
5
716
5743806
CR936600.1
-
5
983
CR936759.1
-
5
683
KU178642.1
-
4
617
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0