Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016582.2 | NP_057666.1 | 5 | 1415 | ||
XM_011545095.2 | XP_011543397.1 | 5 | 1710 | |||
GenBank mRNA | AB020598.1 | - | 5 | 1415 | ||
AF135600.1 | - | 5 | 1308 | |||
AK127216.1 | - | 1 | 2387 | |||
BC037974.1 | - | 5 | 1275 | 5213632 |