Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276380.1 | NP_001263309.1 | 3 | 898 | ||
NM_016649.3 | NP_057733.2 | 3 | 909 | |||
XM_017027874.1 | XP_016883363.1 | 3 | 2361 | |||
GenBank mRNA | AF068285.1 | - | 2 | 266 | ||
AK000375.1 | - | 3 | 910 | |||
AK022369.1 | - | 3 | 860 | |||
AK074299.1 | - | 2 | 395 | |||
BC020952.1 | - | 2 | 528 | |||
BC030513.1 | - | 2 | 859 | |||
BC039706.1 | - | 2 | 850 | |||
BC046107.1 | - | 2 | 848 |