Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016848.5 | NP_058544.3 | 7 | 1221 | ||
XM_011518785.2 | XP_011517087.1 | 7 | 4751 | |||
XM_011518786.2 | XP_011517088.1 | 8 | 600 | |||
XM_017014810.1 | XP_016870299.1 | 8 | 4826 | |||
GenBank mRNA | BC026314.1 | - | 7 | 1222 | 4819086 | |
BX641139.1 | - | 3 | 3674 | |||
D84361.1 | - | 7 | 1094 | |||
KU176125.1 | - | 7 | 518 |