Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017623.4 | NP_060093.3 | 8 | 2263 | ||
NM_199078.2 | NP_951060.1 | 7 | 2119 | |||
XM_017003800.1 | XP_016859289.1 | 8 | 3206 | |||
GenBank mRNA | AF216965.1 | - | 8 | 1362 | ||
AK000025.1 | - | 7 | 1181 | |||
AK126847.1 | - | 8 | 2263 | |||
AK128698.1 | - | 1 | 1103 | |||
BC018604.1 | - | 7 | 1008 | |||
BC022944.1 | - | 5 | 707 | |||
BC037272.1 | - | 8 | 2272 | 4823510 | ||
CR749702.1 | - | 4 | 1761 |