Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075099.1 | NP_001068567.1 | 2 | 224 | ||
NM_003726.3 | NP_003717.3 | 2 | 224 | |||
XM_005257755.3 | XP_005257812.1 | 2 | 283 | |||
XM_017025257.1 | XP_016880746.1 | 2 | 172 | |||
XM_017025259.1 | XP_016880748.1 | 2 | 242 | |||
GenBank mRNA | AK290818.1 | - | 2 | 211 | ||
BC047870.1 | - | 2 | 196 | 5747375 | ||
DR007155.1 | - | 2 | 229 | |||
EU668346.1 | - | 2 | 196 | |||
Y11215.1 | - | 2 | 182 |