Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271028.1 | NP_001257957.1 | 3 | 741 | ||
NM_021153.3 | NP_066976.1 | 3 | 741 | |||
NR_073130.1 | 3 | 741 | ||||
XM_011525932.1 | XP_011524234.1 | 2 | 519 | |||
GenBank mRNA | AJ007607.1 | - | 3 | 582 | ||
AK304000.1 | - | 3 | 691 | |||
AK315257.1 | - | 3 | 743 | |||
AY358654.1 | - | 2 | 480 | |||
BC015877.1 | - | 3 | 633 | 4716394 |