Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018110.3 | NP_060580.2 | 9 | 2366 | ||
XM_005256042.3 | XP_005256099.1 | 7 | 2425 | |||
XM_005256043.2 | XP_005256100.1 | 8 | 2308 | |||
GenBank mRNA | BC001540.2 | - | 8 | 2065 | 2988350 | |
BC003541.1 | - | 9 | 2307 | 3611472 |