Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031412.2 | NP_113600.1 | 4 | 1615 | ||
GenBank mRNA | AF087847.1 | - | 4 | 1656 | ||
AK056846.1 | - | 6 | 1776 | |||
AK092075.1 | - | 4 | 2045 | |||
AK124150.1 | - | 1 | 1814 | |||
AL136676.1 | - | 4 | 1601 | |||
BC009309.2 | - | 4 | 1612 | 4100219 | ||
BC028315.1 | - | 4 | 1615 | 5183078 | ||
BX648781.1 | - | 5 | 2993 |
Gene Symbol: GABARAPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028287.1 | 1 | 1634 | |||
GenBank mRNA | AF180519.1 | - | 1 | 1624 |