Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 9 | 1428 | ||
NM_015574.1 | NP_056389.1 | 9 | 1810 | |||
NM_032217.4 | NP_115593.3 | 9 | 1810 | |||
NM_198889.2 | NP_942592.1 | 9 | 1810 | |||
XM_005265671.3 | XP_005265728.1 | 9 | 2154 | |||
XM_005265672.3 | XP_005265729.1 | 9 | 2154 | |||
XM_005265673.3 | XP_005265730.1 | 9 | 2154 | |||
XM_017008011.1 | XP_016863500.1 | 9 | 1385 | |||
XM_017008012.1 | XP_016863501.1 | 9 | 1385 | |||
XM_017008013.1 | XP_016863502.1 | 9 | 1385 | |||
XM_017008014.1 | XP_016863503.1 | 9 | 2154 | |||
XM_017008015.1 | XP_016863504.1 | 9 | 1385 | |||
XM_017008016.1 | XP_016863505.1 | 9 | 2154 | |||
XM_017008017.1 | XP_016863506.1 | 9 | 1385 | |||
GenBank mRNA | AB014597.2 | - | 9 | 1334 | ||
AF308285.1 | - | 9 | 1797 | |||
AK055209.1 | - | 1 | 38 | |||
AK300302.1 | - | 9 | 1384 | |||
AK307581.1 | - | 9 | 1810 | |||
AL833287.1 | - | 6 | 987 | |||
BC004173.2 | - | 9 | 1791 | |||
BC007747.2 | - | 7 | 1140 | |||
BC043394.1 | - | 9 | 1779 | |||
BX648035.1 | - | 9 | 1383 |