Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145804.2 | NP_665803.2 | 11, 12 | 2833 | ||
GenBank mRNA | AK091560.1 | - | 8, 9 | 1051 | ||
AK095632.1 | - | 11, 12 | 2080 | |||
AK291744.1 | - | 11, 12 | 2834 | |||
BC093968.1 | - | 11, 12 | 1945 | 7939813 | ||
BX640841.1 | - | 3, 4 | 534 |