Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 3 | 428 | ||
NM_024021.3 | NP_076926.2 | 4 | 574 | |||
NM_148975.2 | NP_683876.1 | 3 | 428 | |||
XM_017017909.1 | XP_016873398.1 | 3 | 441 | |||
GenBank mRNA | AB013102.1 | - | 3 | 269 | ||
AB022821.1 | - | 3 | 293 | |||
AF237912.1 | - | 3 | 379 | |||
AF350500.1 | - | 2 | 332 | |||
AF354928.1 | - | 2 | 252 | |||
AK292089.1 | - | 3 | 383 | |||
AK314033.1 | - | 3 | 271 | |||
BC020648.1 | - | 3 | 303 | 4720088 | ||
CB046393.1 | - | 3 | 297 |