Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139488.1 | NP_001132960.1 | 12 | 1966 | ||
NM_015376.2 | NP_056191.1 | 11 | 1521 | |||
NM_170672.2 | NP_733772.1 | 13 | 1879 | |||
XM_005264246.3 | XP_005264303.1 | 12 | 2303 | |||
XM_011532746.2 | XP_011531048.1 | 13 | 1753 | |||
XM_011532747.2 | XP_011531049.1 | 12 | 1768 | |||
XM_011532748.2 | XP_011531050.2 | 12 | 1614 | |||
XM_017003759.1 | XP_016859248.1 | 14 | 3229 | |||
XM_017003760.1 | XP_016859249.1 | 13 | 3701 | |||
XM_017003761.1 | XP_016859250.1 | 12 | 1631 | |||
XM_017003762.1 | XP_016859251.1 | 13 | 1877 | |||
XM_017003763.1 | XP_016859252.1 | 12 | 1607 | |||
GenBank mRNA | AB020653.1 | - | 11 | 1521 | ||
AK124316.1 | - | 2 | 2767 | |||
AK292983.1 | - | 13 | 1790 | |||
BC027849.1 | - | 13 | 1879 | 5203041 | ||
BX647990.1 | - | 12 | 1574 |