Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177424.2 | NP_803173.1 | 9 | 1121 | ||
GenBank mRNA | AF123769.1 | - | 9 | 1068 | ||
AK095898.1 | - | 1 | 1035 | |||
AK297585.1 | - | 9 | 1049 | |||
AK299982.1 | - | 9 | 953 | |||
AK315880.1 | - | 7 | 752 | |||
AL035306.1 | - | 9 | 1064 | |||
BC046999.1 | - | 9 | 1049 | 3851266 |